Learn More
Meso Scale Discovery U-PLEX NHP IL-10 ASY
SDP
Catalog No. 5000081137
Encompass
The Meso Scale Discovery U-PLEX NHP IL-10 Assay is an electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 10 (IL-10) in serum plasma and cell culture supernatants The assay uses MSD s proprietary SECTOR Imager and provides high sensitivity a broad dynamic range and requires low sample volume The lower limit of detection is 0 04 pg/mL The assay can be multiplexed with other U-PLEX biomarkers for efficient analysis The kit includes a 96-well plate coated with capture antibody detection antibody read buffer calibrator diluent wash buffer and a vial of lyophilized calibrator
Catalog No. 50-000-81137 Supplier Meso Scale Discovery Supplier No. K156TZK1
The Meso Scale Discovery U-PLEX NHP IL-10 Assay is an electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 10 (IL-10) in serum plasma and cell culture supernatants The assay uses MSD s proprietary SECTOR Imager and provides high sensitivity a broad dynamic range and requires low sample volume The lower limit of detection is 0 04 pg/mL The assay can be multiplexed with other U-PLEX biomarkers for efficient analysis The kit includes a 96-well plate coated with capture antibody detection antibody read buffer calibrator diluent wash buffer and a vial of lyophilized calibrator