Learn More
Meso Scale Discovery U-PLEX NHP IL-10 ASY384-well
SDP
Catalog No. 5000078989
Encompass
The Meso Scale Discovery U-PLEX NHP IL-10 384-Well Assay is an electrochemiluminescent immunoassay for measuring human interleukin 10 (IL-10) in serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology for sensitive results in 3 5 hours The kit includes a 384-well plate with capture antibody detection antibody with ruthenium label read buffer 10X diluent and calibrators A 25 L sample volume is ideal for precious samples with a dynamic range of 48 - 20 000 pg/mL for various sample types
Catalog No. 50-000-78989 Supplier Meso Scale Discovery Supplier No. K256TZK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX NHP IL-10 384-Well Assay is an electrochemiluminescent immunoassay for measuring human interleukin 10 (IL-10) in serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology for sensitive results in 3 5 hours The kit includes a 384-well plate with capture antibody detection antibody with ruthenium label read buffer 10X diluent and calibrators A 25 L sample volume is ideal for precious samples with a dynamic range of 48 - 20 000 pg/mL for various sample types