Learn More
Meso Scale Discovery U-PLEX NHP IL-12/IL-23p40 ASY
SDP
Catalog No. 5000080126
Encompass
The Meso Scale Discovery U-PLEX NHP IL-12/IL-23p40 Assay is an electrochemiluminescent immunoassay for the quantitative measurement of IL-12/IL-23p40 in non-human primate serum plasma and cell culture supernatants This assay utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible quantitation of IL-12/IL-23p40 with a lower limit of detection of 1 3 pg/mL and an upper limit of quantification of 10 000 pg/mL The assay requires 50 L of sample and can be read on MSD s SECTOR Imager instruments This IL-12/IL-23p40 kit provides components for one 96-well plate including plates calibrators controls detection antibody and buffers
Catalog No. 50-000-80126 Supplier Meso Scale Discovery Supplier No. K156UQK1
The Meso Scale Discovery U-PLEX NHP IL-12/IL-23p40 Assay is an electrochemiluminescent immunoassay for the quantitative measurement of IL-12/IL-23p40 in non-human primate serum plasma and cell culture supernatants This assay utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible quantitation of IL-12/IL-23p40 with a lower limit of detection of 1 3 pg/mL and an upper limit of quantification of 10 000 pg/mL The assay requires 50 L of sample and can be read on MSD s SECTOR Imager instruments This IL-12/IL-23p40 kit provides components for one 96-well plate including plates calibrators controls detection antibody and buffers