Learn More
Meso Scale Discovery U-PLEX NHP IL-12/IL-23p40 ASY
SDP
Catalog No. 5000080141
Encompass
The Meso Scale Discovery U-PLEX NHP IL-12/IL-23p40 Assay is an electrochemiluminescent immunoassay for the quantitative measurement of IL-12/IL-23p40 in non-human primate serum plasma and cell culture supernatants This assay uses MSD s proprietary SECTOR Imager to provide sensitive and accurate detection of IL-12/IL-23p40 levels down to 0 04 pg/mL The assay can be used to monitor immune responses and inflammation in non-human primate models of human disease With a short 2 5 hour protocol and minimal sample volume requirements this assay enables efficient and high-throughput analysis of IL-12/IL-23p40 levels
Catalog No. 50-000-80141 Supplier Meso Scale Discovery Supplier No. K156UQK2
The Meso Scale Discovery U-PLEX NHP IL-12/IL-23p40 Assay is an electrochemiluminescent immunoassay for the quantitative measurement of IL-12/IL-23p40 in non-human primate serum plasma and cell culture supernatants This assay uses MSD s proprietary SECTOR Imager to provide sensitive and accurate detection of IL-12/IL-23p40 levels down to 0 04 pg/mL The assay can be used to monitor immune responses and inflammation in non-human primate models of human disease With a short 2 5 hour protocol and minimal sample volume requirements this assay enables efficient and high-throughput analysis of IL-12/IL-23p40 levels