Learn More
Meso Scale Discovery U-PLEX NHP IL-12p70 ASY
SDP
Catalog No. 5000079266
Encompass
The Meso Scale Discovery U-PLEX NHP IL-12p70 Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human IL-12p70 in serum plasma and cell culture supernatants The assay uses MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results with low sample volume requirements The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies conjugated to ruthenium labels calibrators and reagents required to perform the assay The lower limit of detection is 1 4 pg/mL and the assay range is up to 10 000 pg/mL
This item is not returnable. View return policy
Catalog No. 50-000-79266 Supplier Meso Scale Discovery Supplier No. K156UAK2
The Meso Scale Discovery U-PLEX NHP IL-12p70 Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human IL-12p70 in serum plasma and cell culture supernatants The assay uses MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results with low sample volume requirements The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies conjugated to ruthenium labels calibrators and reagents required to perform the assay The lower limit of detection is 1 4 pg/mL and the assay range is up to 10 000 pg/mL