Learn More
Meso Scale Discovery U-PLEX NHP IL-13 ASY 25 PL
SDP
Catalog No. 5000079963
Encompass
The Meso Scale Discovery U-PLEX NHP IL-13 Assay is an electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 13 (IL-13) in serum plasma and cell culture supernatants The assay uses MSD s proprietary SECTOR Imager and provides high sensitivity a broad dynamic range and requires low sample volume The lower limit of detection is 0 04 pg/mL The assay can be multiplexed with other U-PLEX biomarkers for efficient and customizable biomarker profiling This IL-13 assay delivers rapid results in just 3 5 hours
Catalog No. 50-000-79963 Supplier Meso Scale Discovery Supplier No. K156UBK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX NHP IL-13 Assay is an electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 13 (IL-13) in serum plasma and cell culture supernatants The assay uses MSD s proprietary SECTOR Imager and provides high sensitivity a broad dynamic range and requires low sample volume The lower limit of detection is 0 04 pg/mL The assay can be multiplexed with other U-PLEX biomarkers for efficient and customizable biomarker profiling This IL-13 assay delivers rapid results in just 3 5 hours