Learn More
Meso Scale Discovery U-PLEX NHP IL-13 ASY
SDP
Catalog No. 5000079903
Encompass
The Meso Scale Discovery U-PLEX NHP IL-13 Assay is an electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 13 (IL-13) in serum plasma and cell culture supernatants This assay utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible quantitation of IL-13 with a lower limit of detection of 0 07 pg/mL and an upper limit of quantification of 2500 pg/mL The assay can be multiplexed with other U-PLEX biomarkers and run on MSD s QuickPlex SQ 120 instrument in as little as 3 5 hours This assay allows for the accurate quantitation of IL-13 levels in biological samples
Catalog No. 50-000-79903 Supplier Meso Scale Discovery Supplier No. K156UBK2
The Meso Scale Discovery U-PLEX NHP IL-13 Assay is an electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 13 (IL-13) in serum plasma and cell culture supernatants This assay utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible quantitation of IL-13 with a lower limit of detection of 0 07 pg/mL and an upper limit of quantification of 2500 pg/mL The assay can be multiplexed with other U-PLEX biomarkers and run on MSD s QuickPlex SQ 120 instrument in as little as 3 5 hours This assay allows for the accurate quantitation of IL-13 levels in biological samples