Learn More
Meso Scale Discovery U-PLEX NHP IL-13 ASY
SDP
Catalog No. 5000080848
Encompass
The Meso Scale Discovery U-PLEX NHP IL-13 Assay Kit is a sensitive immunoassay for quantifying natural human IL-13 in serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection technology offering high sensitivity a wide dynamic range and a low detection limit Key features include a 2 5-hour protocol minimal sample volume and a reusable plate format making it an eco-friendly and cost-effective solution for accurate IL-13 detection in various human samples
Catalog No. 50-000-80848 Supplier Meso Scale Discovery Supplier No. K156UBK1
The Meso Scale Discovery U-PLEX NHP IL-13 Assay Kit is a sensitive immunoassay for quantifying natural human IL-13 in serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection technology offering high sensitivity a wide dynamic range and a low detection limit Key features include a 2 5-hour protocol minimal sample volume and a reusable plate format making it an eco-friendly and cost-effective solution for accurate IL-13 detection in various human samples