Learn More
Meso Scale Discovery U-PLEX NHP IL-13 ASY384-well
SDP
Catalog No. 5000080008
Encompass
The Meso Scale Discovery U-PLEX NHP IL-13 384-Well Assay Kit is a multiplex immunoassay for measuring human IL-13 in serum plasma and cell culture supernatants It uses electrochemiluminescence detection for high sensitivity and rapid results The kit includes a 384-well plate with capture antibodies detection antibodies and necessary calibrators and controls It analyzes up to 80 samples simultaneously with small volumes offering high performance and reliability for biomarker research and drug discovery
Catalog No. 50-000-80008 Supplier Meso Scale Discovery Supplier No. K256UBK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX NHP IL-13 384-Well Assay Kit is a multiplex immunoassay for measuring human IL-13 in serum plasma and cell culture supernatants It uses electrochemiluminescence detection for high sensitivity and rapid results The kit includes a 384-well plate with capture antibodies detection antibodies and necessary calibrators and controls It analyzes up to 80 samples simultaneously with small volumes offering high performance and reliability for biomarker research and drug discovery