Learn More
Meso Scale Discovery U-PLEX NHP IL-13 ASYQP
SDP
Catalog No. 5000078549
Encompass
The MSD U-PLEX IL-13 Assay is a singleplex electrochemiluminescence immunoassay designed for quantitative measurement of interleukin-13 in non-human primate samples This research-grade assay utilizes Meso Scale Discovery s proprietary U-PLEX platform technology offering high sensitivity and specificity for IL-13 detection The assay is validated for serum plasma and cell culture supernatant samples from non-human primates making it ideal for preclinical research applications studying inflammatory responses and immune system function
Catalog No. 50-000-78549 Supplier Meso Scale Discovery Supplier No. K156UBK22
Add to Cart
Add to Cart
The MSD U-PLEX IL-13 Assay is a singleplex electrochemiluminescence immunoassay designed for quantitative measurement of interleukin-13 in non-human primate samples This research-grade assay utilizes Meso Scale Discovery s proprietary U-PLEX platform technology offering high sensitivity and specificity for IL-13 detection The assay is validated for serum plasma and cell culture supernatant samples from non-human primates making it ideal for preclinical research applications studying inflammatory responses and immune system function