Learn More
Meso Scale Discovery U-PLEX NHP IL-15 ASY 25 PL
SDP
Catalog No. 5000080118
Encompass
The Meso Scale Discovery U-PLEX NHP IL-15 Assay Kit is a multiplex immunoassay that measures natural human IL-15 in serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and low sample consumption The assay requires 50 L of sample and can be read on the MESO QuickPlex SQ 120 instrument The kit includes a plate coated with capture antibodies detection antibodies Read Buffer Sample Diluent and calibrators ranging from 3 2 - 10 000 pg/mL This assay allows for the accurate quantitation of IL-15 levels in human samples
Catalog No. 50-000-80118 Supplier Meso Scale Discovery Supplier No. K156URK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX NHP IL-15 Assay Kit is a multiplex immunoassay that measures natural human IL-15 in serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and low sample consumption The assay requires 50 L of sample and can be read on the MESO QuickPlex SQ 120 instrument The kit includes a plate coated with capture antibodies detection antibodies Read Buffer Sample Diluent and calibrators ranging from 3 2 - 10 000 pg/mL This assay allows for the accurate quantitation of IL-15 levels in human samples