Learn More
Meso Scale Discovery U-PLEX NHP IL-15 ASY
SDP
Catalog No. 5000080137
Encompass
The Meso Scale Discovery U-PLEX NHP IL-15 Assay Kit is a multiplex immunoassay that measures natural human IL-15 in serum plasma and cell culture supernatants This assay uses MSD s electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies calibrators and controls for generating a standard curve and measuring IL-15 levels between 0 06-1500 pg/mL This quantitative immunoassay allows sensitive reproducible and rapid measurement of IL-15
Catalog No. 50-000-80137 Supplier Meso Scale Discovery Supplier No. K156URK1
The Meso Scale Discovery U-PLEX NHP IL-15 Assay Kit is a multiplex immunoassay that measures natural human IL-15 in serum plasma and cell culture supernatants This assay uses MSD s electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies calibrators and controls for generating a standard curve and measuring IL-15 levels between 0 06-1500 pg/mL This quantitative immunoassay allows sensitive reproducible and rapid measurement of IL-15