Learn More
Meso Scale Discovery U-PLEX NHP IL-15 ASY384-well
SDP
Catalog No. 5000079169
Encompass
The Meso Scale Discovery U-PLEX NHP IL-15 384-Well Assay Kit is a multiplex immunoassay for measuring human IL-15 in serum plasma and cell culture supernatants It uses electrochemiluminescence detection for high sensitivity and rapid results The kit includes a 384-well plate with capture antibodies detection antibodies calibrators and controls allowing IL-15 measurement from 25 L samples Detection limits range from 0 06 to 2500 pg/mL An MSD instrument is required The kit lasts 9 months when stored at 2-8 C
Catalog No. 50-000-79169 Supplier Meso Scale Discovery Supplier No. K256URK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX NHP IL-15 384-Well Assay Kit is a multiplex immunoassay for measuring human IL-15 in serum plasma and cell culture supernatants It uses electrochemiluminescence detection for high sensitivity and rapid results The kit includes a 384-well plate with capture antibodies detection antibodies calibrators and controls allowing IL-15 measurement from 25 L samples Detection limits range from 0 06 to 2500 pg/mL An MSD instrument is required The kit lasts 9 months when stored at 2-8 C