Learn More
Meso Scale Discovery U-PLEX NHP IL-15 ASYQP 25 PL
SDP
Catalog No. 5000080707
Encompass
The U-PLEX NHP IL-15 Assay (K156URK) is a specialized immunoassay kit designed for quantitative measurement of Interleukin-15 in non-human primate samples This electrochemiluminescence-based assay utilizes Meso Scale Discovery s U-PLEX platform technology offering high sensitivity and specificity for IL-15 detection in serum plasma and cell culture supernatants The kit provides researchers with reliable tools for cytokine analysis in preclinical studies involving non-human primate models
Catalog No. 50-000-80707 Supplier Meso Scale Discovery Supplier No. K156URK24
Add to Cart
Add to Cart
Request a Quote
The U-PLEX NHP IL-15 Assay (K156URK) is a specialized immunoassay kit designed for quantitative measurement of Interleukin-15 in non-human primate samples This electrochemiluminescence-based assay utilizes Meso Scale Discovery s U-PLEX platform technology offering high sensitivity and specificity for IL-15 detection in serum plasma and cell culture supernatants The kit provides researchers with reliable tools for cytokine analysis in preclinical studies involving non-human primate models