Learn More
Meso Scale Discovery U-PLEX NHP IL-16 ASY 25 PL
SDP
Catalog No. 5000080678
Encompass
The Meso Scale Discovery U-PLEX NHP IL-16 Assay is a multiplex immunoassay kit for measuring human interleukin 16 (IL-16) in serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology for sensitive reproducible results with low sample volume The kit includes a 96-well plate with capture antibodies detection antibodies with SULFO-TAG calibrators controls buffers and seals It detects IL-16 in the range of 1 53-2500 pg/mL offering high sensitivity accuracy and reproducibility for research
Catalog No. 50-000-80678 Supplier Meso Scale Discovery Supplier No. K156USK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX NHP IL-16 Assay is a multiplex immunoassay kit for measuring human interleukin 16 (IL-16) in serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology for sensitive reproducible results with low sample volume The kit includes a 96-well plate with capture antibodies detection antibodies with SULFO-TAG calibrators controls buffers and seals It detects IL-16 in the range of 1 53-2500 pg/mL offering high sensitivity accuracy and reproducibility for research