Learn More
Meso Scale Discovery U-PLEX NHP IL-16 ASY
SDP
Catalog No. 5000080031
Encompass
The Meso Scale Discovery U-PLEX NHP IL-16 Assay is a multiplex immunoassay kit for measuring human interleukin 16 (IL-16) in serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection technology in a 96-well plate format The kit includes capture and detection antibodies calibrator solutions and necessary buffers Sensitivity is 0 3 pg/mL with a range of 0 3-2500 pg/mL Total assay time is about 3 5 hours It aids in cytokine profiling and research on inflammatory and autoimmune conditions
Catalog No. 50-000-80031 Supplier Meso Scale Discovery Supplier No. K156USK2
The Meso Scale Discovery U-PLEX NHP IL-16 Assay is a multiplex immunoassay kit for measuring human interleukin 16 (IL-16) in serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection technology in a 96-well plate format The kit includes capture and detection antibodies calibrator solutions and necessary buffers Sensitivity is 0 3 pg/mL with a range of 0 3-2500 pg/mL Total assay time is about 3 5 hours It aids in cytokine profiling and research on inflammatory and autoimmune conditions