Learn More
Meso Scale Discovery U-PLEX NHP IL-16 ASY
SDP
Catalog No. 5000080757
Encompass
The Meso Scale Discovery U-PLEX NHP IL-16 Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of human interleukin 16 (IL-16) in serum plasma and cell culture supernatants This assay utilizes MSD s proprietary electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies calibrators and controls for generating a standard curve and measuring IL-16 levels This assay allows for rapid reproducible quantitation of IL-16 which plays a role in inflammatory and autoimmune conditions
Catalog No. 50-000-80757 Supplier Meso Scale Discovery Supplier No. K156USK1
Add to Cart
Add to Cart
The Meso Scale Discovery U-PLEX NHP IL-16 Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of human interleukin 16 (IL-16) in serum plasma and cell culture supernatants This assay utilizes MSD s proprietary electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies calibrators and controls for generating a standard curve and measuring IL-16 levels This assay allows for rapid reproducible quantitation of IL-16 which plays a role in inflammatory and autoimmune conditions