Learn More
Meso Scale Discovery U-PLEX NHP IL-16 ASY384-well
SDP
Catalog No. 5000081743
Encompass
The Meso Scale Discovery U-PLEX NHP IL-16 384-Well Assay Kit is a multiplex immunoassay that measures human interleukin 16 (IL-16) in serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample consumption The kit includes a 384-well plate coated with capture antibodies detection antibodies labeled with an electrochemiluminescent compound and calibrator and control solutions It allows simultaneous measurement of IL-16 along with other biomarkers The kit has high performance and reliability for biomarker quantification in biomedical research
Catalog No. 50-000-81743 Supplier Meso Scale Discovery Supplier No. K256USK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX NHP IL-16 384-Well Assay Kit is a multiplex immunoassay that measures human interleukin 16 (IL-16) in serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample consumption The kit includes a 384-well plate coated with capture antibodies detection antibodies labeled with an electrochemiluminescent compound and calibrator and control solutions It allows simultaneous measurement of IL-16 along with other biomarkers The kit has high performance and reliability for biomarker quantification in biomedical research