Learn More
Meso Scale Discovery U-PLEX NHP IL-16 ASYQP 25 PL
SDP
Catalog No. 5000080754
Encompass
Meso Scale Discovery offers IL-16 (Interleukin-16) immunoassay products for research applications IL-16 is a pro-inflammatory cytokine that functions as a chemoattractant for CD4 T cells monocytes and eosinophils MSD s electrochemiluminescence-based detection platform provides sensitive quantitative measurement of IL-16 levels in biological samples These assays are designed for researchers studying inflammatory processes immune cell migration and cytokine signaling pathways in various disease models and clinical research applications
Catalog No. 50-000-80754 Supplier Meso Scale Discovery Supplier No. K156USK24
Add to Cart
Add to Cart
Request a Quote
Meso Scale Discovery offers IL-16 (Interleukin-16) immunoassay products for research applications IL-16 is a pro-inflammatory cytokine that functions as a chemoattractant for CD4 T cells monocytes and eosinophils MSD s electrochemiluminescence-based detection platform provides sensitive quantitative measurement of IL-16 levels in biological samples These assays are designed for researchers studying inflammatory processes immune cell migration and cytokine signaling pathways in various disease models and clinical research applications