Learn More
Meso Scale Discovery U-PLEX NHP IL-17A ASY 25 PL
SDP
Catalog No. 5000080045
Encompass
The Meso Scale Discovery U-PLEX NHP IL-17A Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of IL-17A in non-human primate serum plasma and cell culture supernatants The assay uses MSD s proprietary SECTOR Imager and provides a sensitive and reproducible method for measuring IL-17A with a detection range of 0 05-1000 pg/mL and an LLOQ of 0 05 pg/mL The kit includes a 96-well plate coated with capture antibody detection antibody read buffer calibrator diluent wash buffer and a vial of lyophilized calibrator The assay takes approximately 3 5 hours to complete
Catalog No. 50-000-80045 Supplier Meso Scale Discovery Supplier No. K156UTK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX NHP IL-17A Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of IL-17A in non-human primate serum plasma and cell culture supernatants The assay uses MSD s proprietary SECTOR Imager and provides a sensitive and reproducible method for measuring IL-17A with a detection range of 0 05-1000 pg/mL and an LLOQ of 0 05 pg/mL The kit includes a 96-well plate coated with capture antibody detection antibody read buffer calibrator diluent wash buffer and a vial of lyophilized calibrator The assay takes approximately 3 5 hours to complete