Learn More
Meso Scale Discovery U-PLEX NHP IL-17A ASY
SDP
Catalog No. 5000080134
Encompass
The Meso Scale Discovery U-PLEX NHP IL-17A Assay is an electrochemiluminescent immunoassay for measuring human IL-17A in serum plasma and cell culture supernatants It uses MSD s SECTOR Imager offering high sensitivity broad dynamic range and low sample volume The assay specifically detects natural and recombinant human IL-17A without cross-reactivity to other cytokines It includes a capture antibody-coated plate detection antibody Read Buffer Diluent 2 Calibrator Diluent and lyophilized U-PLEX Calibrator Blend Sensitivity is 0 04 pg/mL with an upper quantification limit of 2500 pg/mL
Catalog No. 50-000-80134 Supplier Meso Scale Discovery Supplier No. K156UTK2
The Meso Scale Discovery U-PLEX NHP IL-17A Assay is an electrochemiluminescent immunoassay for measuring human IL-17A in serum plasma and cell culture supernatants It uses MSD s SECTOR Imager offering high sensitivity broad dynamic range and low sample volume The assay specifically detects natural and recombinant human IL-17A without cross-reactivity to other cytokines It includes a capture antibody-coated plate detection antibody Read Buffer Diluent 2 Calibrator Diluent and lyophilized U-PLEX Calibrator Blend Sensitivity is 0 04 pg/mL with an upper quantification limit of 2500 pg/mL