Learn More
Meso Scale Discovery U-PLEX NHP IL-17A ASYQP
SDP
Catalog No. 5000080115
Encompass
The MSD U-PLEX IL-17A Singleplex Assay is a specialized immunoassay designed for measuring interleukin-17A levels in non-human primate samples This electrochemiluminescence-based platform offers high sensitivity and precision for cytokine detection in research applications The singleplex format allows focused analysis of IL-17A a key inflammatory cytokine involved in immune responses and autoimmune diseases Compatible with various sample types this assay provides reliable quantification for preclinical studies and biomedical research requiring accurate IL-17A measurement in primate models
Catalog No. 50-000-80115 Supplier Meso Scale Discovery Supplier No. K156UTK21
The MSD U-PLEX IL-17A Singleplex Assay is a specialized immunoassay designed for measuring interleukin-17A levels in non-human primate samples This electrochemiluminescence-based platform offers high sensitivity and precision for cytokine detection in research applications The singleplex format allows focused analysis of IL-17A a key inflammatory cytokine involved in immune responses and autoimmune diseases Compatible with various sample types this assay provides reliable quantification for preclinical studies and biomedical research requiring accurate IL-17A measurement in primate models