Learn More
Meso Scale Discovery U-PLEX NHP IL-17A/F ASY
SDP
Catalog No. 5000080318
Encompass
The Meso Scale Discovery U-PLEX NHP IL-17A/F Assay is a multiplex immunoassay kit that enables simultaneous quantification of human IL-17A and IL-17F in serum plasma and cell culture supernatants The assay utilizes MSD s proprietary electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements This assay allows for the accurate measurement of IL-17A and IL-17F which are inflammatory cytokines involved in autoimmune diseases With minimal hands-on time this kit serves as a useful tool for biomarker research and immunology studies
Catalog No. 50-000-80318 Supplier Meso Scale Discovery Supplier No. K156VYK2
The Meso Scale Discovery U-PLEX NHP IL-17A/F Assay is a multiplex immunoassay kit that enables simultaneous quantification of human IL-17A and IL-17F in serum plasma and cell culture supernatants The assay utilizes MSD s proprietary electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements This assay allows for the accurate measurement of IL-17A and IL-17F which are inflammatory cytokines involved in autoimmune diseases With minimal hands-on time this kit serves as a useful tool for biomarker research and immunology studies