Learn More
Meso Scale Discovery U-PLEX NHP IL-17A/F ASYQP
SDP
Catalog No. 5000080130
Encompass
MSD s IL-17A/F assay is an electrochemiluminescence-based immunoassay for quantifying IL-17A/F heterodimer cytokine levels in biological samples This multiplex platform enables sensitive detection and measurement of this inflammatory biomarker commonly used in research applications studying immune responses autoimmune diseases and inflammatory conditions The assay provides reliable reproducible results for laboratory research purposes
Catalog No. 50-000-80130 Supplier Meso Scale Discovery Supplier No. K156VYK22
MSD s IL-17A/F assay is an electrochemiluminescence-based immunoassay for quantifying IL-17A/F heterodimer cytokine levels in biological samples This multiplex platform enables sensitive detection and measurement of this inflammatory biomarker commonly used in research applications studying immune responses autoimmune diseases and inflammatory conditions The assay provides reliable reproducible results for laboratory research purposes