Learn More
Meso Scale Discovery U-PLEX NHP IL-17B ASY
SDP
Catalog No. 5000078628
Encompass
The Meso Scale Discovery U-PLEX NHP IL-17B Assay is a multiplex immunoassay kit for measuring IL-17B in non-human primate serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection in a 96-well plate format requiring only 25 L per well The kit includes capture and detection antibodies calibrators and diluent It is compatible with MSD SECTOR Imager instruments and analyzed via MSD Discovery Workbench software offering a fast reproducible method for immune monitoring and biomarker studies in non-human primates
Catalog No. 50-000-78628 Supplier Meso Scale Discovery Supplier No. K156XNK1
The Meso Scale Discovery U-PLEX NHP IL-17B Assay is a multiplex immunoassay kit for measuring IL-17B in non-human primate serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection in a 96-well plate format requiring only 25 L per well The kit includes capture and detection antibodies calibrators and diluent It is compatible with MSD SECTOR Imager instruments and analyzed via MSD Discovery Workbench software offering a fast reproducible method for immune monitoring and biomarker studies in non-human primates