Learn More
Meso Scale Discovery U-PLEX NHP IL-17C ASY 25 PL
SDP
Catalog No. 5000080061
Encompass
The Meso Scale Discovery U-PLEX NHP IL-17C Assay is a sensitive immunoassay for measuring natural and recombinant non-human primate interleukin 17C (IL-17C) in serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection technology offering high sensitivity a wide dynamic range and compatibility with common sample matrices With detection limits from 0 04 pg/mL to 10 000 pg/mL it accurately quantifies IL-17C The U-PLEX format allows rapid testing and minimal sample use in a 96-well plate
Catalog No. 50-000-80061 Supplier Meso Scale Discovery Supplier No. K156WJK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX NHP IL-17C Assay is a sensitive immunoassay for measuring natural and recombinant non-human primate interleukin 17C (IL-17C) in serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection technology offering high sensitivity a wide dynamic range and compatibility with common sample matrices With detection limits from 0 04 pg/mL to 10 000 pg/mL it accurately quantifies IL-17C The U-PLEX format allows rapid testing and minimal sample use in a 96-well plate