Learn More
Meso Scale Discovery U-PLEX NHP IL-17C ASY
SDP
Catalog No. 5000080106
Encompass
The Meso Scale Discovery U-PLEX NHP IL-17C Assay is a multiplex immunoassay kit for the quantitative measurement of IL-17C in non-human primate serum plasma and cell culture supernatants The assay uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and low sample consumption The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent tags calibrator diluent wash buffer read buffer and a 10-point IL-17C calibration curve The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 10 000 pg/mL
Catalog No. 50-000-80106 Supplier Meso Scale Discovery Supplier No. K156WJK1
The Meso Scale Discovery U-PLEX NHP IL-17C Assay is a multiplex immunoassay kit for the quantitative measurement of IL-17C in non-human primate serum plasma and cell culture supernatants The assay uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and low sample consumption The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent tags calibrator diluent wash buffer read buffer and a 10-point IL-17C calibration curve The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 10 000 pg/mL