Learn More
Meso Scale Discovery U-PLEX NHP IL-17C ASY384-well
SDP
Catalog No. 5000079204
Encompass
The Meso Scale Discovery U-PLEX NHP IL-17C 384-Well Assay is a multiplex immunoassay kit that enables the simultaneous quantitative measurement of human IL-17C in serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides a 384-well plate formatted assay complete with analyte-specific antibodies pre-coated onto distinct spots This assay kit offers high sensitivity broad dynamic range and requires low sample volume It can be used for applications such as biomarker discovery and validation With a testing time of just 3 5 hours this kit enables rapid quantification of IL-17C
Catalog No. 50-000-79204 Supplier Meso Scale Discovery Supplier No. K256WJK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX NHP IL-17C 384-Well Assay is a multiplex immunoassay kit that enables the simultaneous quantitative measurement of human IL-17C in serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides a 384-well plate formatted assay complete with analyte-specific antibodies pre-coated onto distinct spots This assay kit offers high sensitivity broad dynamic range and requires low sample volume It can be used for applications such as biomarker discovery and validation With a testing time of just 3 5 hours this kit enables rapid quantification of IL-17C