Learn More
Meso Scale Discovery U-PLEX NHP IL-17D ASY
SDP
Catalog No. 5000078464
Encompass
The Meso Scale Discovery U-PLEX NHP IL-17D Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of IL-17D in non-human primate (NHP) samples such as serum plasma and cell culture supernatants This assay utilizes MSD s proprietary electrochemiluminescent detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements The kit includes a 96-well plate pre-coated with capture antibodies for IL-17D detection antibodies calibrators controls 10X diluent read buffer and plate seals The assay can detect IL-17D concentrations as low as 61 pg/mL with an approximate 3 5 hour total assay time
Catalog No. 50-000-78464 Supplier Meso Scale Discovery Supplier No. K156XOK2
Add to Cart
Add to Cart
The Meso Scale Discovery U-PLEX NHP IL-17D Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of IL-17D in non-human primate (NHP) samples such as serum plasma and cell culture supernatants This assay utilizes MSD s proprietary electrochemiluminescent detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements The kit includes a 96-well plate pre-coated with capture antibodies for IL-17D detection antibodies calibrators controls 10X diluent read buffer and plate seals The assay can detect IL-17D concentrations as low as 61 pg/mL with an approximate 3 5 hour total assay time