Learn More
Meso Scale Discovery U-PLEX NHP IL-17D ASY
SDP
Catalog No. 5000080780
Encompass
The Meso Scale Discovery U-PLEX NHP IL-17D Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of human and non-human primate interleukin 17D (IL-17D) This assay utilizes MSD s proprietary electrochemiluminescent detection technology and is provided in a convenient 96-well plate format The kit includes all necessary reagents and diluents to perform the assay and can accurately quantify IL-17D concentrations with high sensitivity and wide dynamic range This product would be useful for researchers studying immune system function and inflammation
Catalog No. 50-000-80780 Supplier Meso Scale Discovery Supplier No. K156XOK1
The Meso Scale Discovery U-PLEX NHP IL-17D Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of human and non-human primate interleukin 17D (IL-17D) This assay utilizes MSD s proprietary electrochemiluminescent detection technology and is provided in a convenient 96-well plate format The kit includes all necessary reagents and diluents to perform the assay and can accurately quantify IL-17D concentrations with high sensitivity and wide dynamic range This product would be useful for researchers studying immune system function and inflammation