Learn More
Meso Scale Discovery U-PLEX NHP IL-17D ASY384-well
SDP
Catalog No. 5000079206
Encompass
The Meso Scale Discovery U-PLEX NHP IL-17D 384-Well Assay is a multiplex immunoassay panel that measures natural human IL-17D levels in serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology in a 384-well plate format providing high sensitivity a wide dynamic range and low sample consumption The assay requires 10 L of sample per well and can perform up to 40 tests across two plates It includes a plate coated with capture antibodies detection antibodies and calibrator and control solutions The assay has a typical lower limit of detection of 0 04 pg/mL and an upper limit of quantification of 2500 pg/mL for IL-17D
Catalog No. 50-000-79206 Supplier Meso Scale Discovery Supplier No. K256XOK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX NHP IL-17D 384-Well Assay is a multiplex immunoassay panel that measures natural human IL-17D levels in serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology in a 384-well plate format providing high sensitivity a wide dynamic range and low sample consumption The assay requires 10 L of sample per well and can perform up to 40 tests across two plates It includes a plate coated with capture antibodies detection antibodies and calibrator and control solutions The assay has a typical lower limit of detection of 0 04 pg/mL and an upper limit of quantification of 2500 pg/mL for IL-17D