Learn More
Meso Scale Discovery U-PLEX NHP IL-17F ASY 25 PL
SDP
Catalog No. 5000080673
Encompass
The Meso Scale Discovery U-PLEX NHP IL-17F Assay is a multiplex immunoassay kit for measuring human and non-human primate IL-17F It uses MSD s electrochemiluminescent detection in a 96-well plate format The kit includes capture and detection antibodies and calibrator solutions for standard curves It offers high sensitivity low sample volume and a 3 5-hour protocol This assay aids in understanding IL-17F s role in autoimmunity inflammation and immune-mediated diseases
Catalog No. 50-000-80673 Supplier Meso Scale Discovery Supplier No. K156WAK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX NHP IL-17F Assay is a multiplex immunoassay kit for measuring human and non-human primate IL-17F It uses MSD s electrochemiluminescent detection in a 96-well plate format The kit includes capture and detection antibodies and calibrator solutions for standard curves It offers high sensitivity low sample volume and a 3 5-hour protocol This assay aids in understanding IL-17F s role in autoimmunity inflammation and immune-mediated diseases