Learn More
Meso Scale Discovery U-PLEX NHP IL-17F ASY384-well
SDP
Catalog No. 5000078424
Encompass
The Meso Scale Discovery U-PLEX NHP IL-17F 384-Well Assay Kit is a multiplex immunoassay for measuring human IL-17F in serum plasma and cell culture supernatants It uses electrochemiluminescence detection for high sensitivity and precision The kit includes a plate with capture antibodies detection antibodies with electrochemiluminescent labels calibrators and diluent It quantifies IL-17F from 15 6 to 1000 pg/mL with a detection limit of 4 2 pg/mL suitable for life science research
Catalog No. 50-000-78424 Supplier Meso Scale Discovery Supplier No. K256WAK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX NHP IL-17F 384-Well Assay Kit is a multiplex immunoassay for measuring human IL-17F in serum plasma and cell culture supernatants It uses electrochemiluminescence detection for high sensitivity and precision The kit includes a plate with capture antibodies detection antibodies with electrochemiluminescent labels calibrators and diluent It quantifies IL-17F from 15 6 to 1000 pg/mL with a detection limit of 4 2 pg/mL suitable for life science research