Learn More
Meso Scale Discovery U-PLEX NHP IL-18 ASY 25 PL
SDP
Catalog No. 5000080129
Encompass
The Meso Scale Discovery U-PLEX NHP IL-18 Assay is an electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 18 (IL-18) in serum plasma and cell culture supernatants This assay utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results with low sample volume requirements The assay has a detection range of 19 5-20 000 pg/mL and includes a plate coated with capture antibodies detection antibodies labeled with ruthenium read buffer wash buffer blocker solutions calibrator diluent and a calibrator This assay allows for the accurate quantitation of IL-18 levels in human samples
Catalog No. 50-000-80129 Supplier Meso Scale Discovery Supplier No. K156VJK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX NHP IL-18 Assay is an electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 18 (IL-18) in serum plasma and cell culture supernatants This assay utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results with low sample volume requirements The assay has a detection range of 19 5-20 000 pg/mL and includes a plate coated with capture antibodies detection antibodies labeled with ruthenium read buffer wash buffer blocker solutions calibrator diluent and a calibrator This assay allows for the accurate quantitation of IL-18 levels in human samples