Learn More
Meso Scale Discovery U-PLEX NHP IL-18 ASY
SDP
Catalog No. 5000079900
Encompass
The Meso Scale Discovery U-PLEX NHP IL-18 Assay is an electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 18 (IL-18) in serum plasma and cell culture supernatants This assay utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible quantitation of IL-18 with a lower limit of detection of 0 91 pg/mL and an upper limit of quantification of 1500 pg/mL The assay requires a small sample volume of 50 L and has a total assay time of approximately 3 5 hours This product can aid in understanding IL-18 associated inflammatory diseases
Catalog No. 50-000-79900 Supplier Meso Scale Discovery Supplier No. K156VJK1
The Meso Scale Discovery U-PLEX NHP IL-18 Assay is an electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 18 (IL-18) in serum plasma and cell culture supernatants This assay utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible quantitation of IL-18 with a lower limit of detection of 0 91 pg/mL and an upper limit of quantification of 1500 pg/mL The assay requires a small sample volume of 50 L and has a total assay time of approximately 3 5 hours This product can aid in understanding IL-18 associated inflammatory diseases