Learn More
Meso Scale Discovery U-PLEX NHP IL-18 ASY
SDP
Catalog No. 5000080101
Encompass
The Meso Scale Discovery U-PLEX NHP IL-18 Assay is an electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 18 (IL-18) in serum plasma and cell culture supernatants This assay utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results in just 3 5 hours with a lower limit of detection of 0 91 pg/mL and an upper limit of quantification of 2500 pg/mL The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to ruthenium label calibrators and assay diluent This assay allows for high-throughput multiplexed cytokine profiling from small sample volumes
Catalog No. 50-000-80101 Supplier Meso Scale Discovery Supplier No. K156VJK2
Add to Cart
Add to Cart
The Meso Scale Discovery U-PLEX NHP IL-18 Assay is an electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 18 (IL-18) in serum plasma and cell culture supernatants This assay utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results in just 3 5 hours with a lower limit of detection of 0 91 pg/mL and an upper limit of quantification of 2500 pg/mL The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to ruthenium label calibrators and assay diluent This assay allows for high-throughput multiplexed cytokine profiling from small sample volumes