Learn More
Meso Scale Discovery U-PLEX NHP IL-18 ASY384-well
SDP
Catalog No. 5000080222
Encompass
The Meso Scale Discovery U-PLEX NHP IL-18 384-Well Assay is a multiplex immunoassay kit that enables the simultaneous quantitative measurement of human interleukin 18 (IL-18) in serum plasma and cell culture supernatants It utilizes MSD s electrochemiluminescent detection technology and provides a 384-well plate formatted assay with a testing range of 0 61-2500 pg/mL The assay requires a low sample volume of 25 L and has high sensitivity and broad dynamic range This product can aid in understanding IL-18 s role in inflammation and autoimmune diseases
Catalog No. 50-000-80222 Supplier Meso Scale Discovery Supplier No. K256VJK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX NHP IL-18 384-Well Assay is a multiplex immunoassay kit that enables the simultaneous quantitative measurement of human interleukin 18 (IL-18) in serum plasma and cell culture supernatants It utilizes MSD s electrochemiluminescent detection technology and provides a 384-well plate formatted assay with a testing range of 0 61-2500 pg/mL The assay requires a low sample volume of 25 L and has high sensitivity and broad dynamic range This product can aid in understanding IL-18 s role in inflammation and autoimmune diseases