Learn More
Meso Scale Discovery U-PLEX NHP IL-1B ASY
SDP
Catalog No. 5000078547
Encompass
The Meso Scale Discovery U-PLEX NHP IL-1beta Assay is an electrochemiluminescent immunoassay for the quantitative measurement of IL-1beta in non-human primate serum plasma and cell culture supernatants It utilizes MSD s proprietary SECTOR Imager and provides superior sensitivity a broad dynamic range and requires low sample volume The assay has high specificity for non-human primate IL-1beta with no cross-reactivity to other cytokines It includes a plate coated with capture antibody detection antibody Read Buffer Diluent 2 Calibrator Diluent and lyophilized calibrator The assay has a typical sensitivity of 0 04 pg/mL and an upper limit of quantitation of 1500 pg/mL
Catalog No. 50-000-78547 Supplier Meso Scale Discovery Supplier No. K156TUK2
The Meso Scale Discovery U-PLEX NHP IL-1beta Assay is an electrochemiluminescent immunoassay for the quantitative measurement of IL-1beta in non-human primate serum plasma and cell culture supernatants It utilizes MSD s proprietary SECTOR Imager and provides superior sensitivity a broad dynamic range and requires low sample volume The assay has high specificity for non-human primate IL-1beta with no cross-reactivity to other cytokines It includes a plate coated with capture antibody detection antibody Read Buffer Diluent 2 Calibrator Diluent and lyophilized calibrator The assay has a typical sensitivity of 0 04 pg/mL and an upper limit of quantitation of 1500 pg/mL