Learn More
Meso Scale Discovery U-PLEX NHP IL-1B ASY384-well
SDP
Catalog No. 5000081223
Encompass
The Meso Scale Discovery U-PLEX NHP IL-1beta 384-well Assay Kit is a multiplex immunoassay that measures human IL-1beta in serum plasma and cell culture supernatants It uses electrochemiluminescence detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample consumption The kit includes a 384-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent labels calibrators and diluent It allows simultaneous measurement of IL-1beta along with other biomarkers The kit has high performance and is designed for use on the MESO QuickPlex SQ 120 instrument
Catalog No. 50-000-81223 Supplier Meso Scale Discovery Supplier No. K256TUK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX NHP IL-1beta 384-well Assay Kit is a multiplex immunoassay that measures human IL-1beta in serum plasma and cell culture supernatants It uses electrochemiluminescence detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample consumption The kit includes a 384-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent labels calibrators and diluent It allows simultaneous measurement of IL-1beta along with other biomarkers The kit has high performance and is designed for use on the MESO QuickPlex SQ 120 instrument