Learn More
Meso Scale Discovery U-PLEX NHP IL-1B ASYQP
SDP
Catalog No. 5000078995
Encompass
The U-PLEX NHP IL-1 Assay (K156TUK) is a specialized immunoassay kit designed for measuring interleukin-1 beta levels in non-human primate samples This electrochemiluminescence-based assay provides high sensitivity and specificity for IL-1 detection in research applications The kit utilizes MSD s U-PLEX platform technology offering researchers a reliable tool for cytokine analysis in preclinical studies involving non-human primate models
Catalog No. 50-000-78995 Supplier Meso Scale Discovery Supplier No. K156TUK22
The U-PLEX NHP IL-1 Assay (K156TUK) is a specialized immunoassay kit designed for measuring interleukin-1 beta levels in non-human primate samples This electrochemiluminescence-based assay provides high sensitivity and specificity for IL-1 detection in research applications The kit utilizes MSD s U-PLEX platform technology offering researchers a reliable tool for cytokine analysis in preclinical studies involving non-human primate models