Learn More
Meso Scale Discovery U-PLEX NHP IL-1RA ASY 25 PL
SDP
Catalog No. 5000081073
Encompass
The Meso Scale Discovery U-PLEX NHP IL-1ra Assay is a multiplex immunoassay kit for measuring human interleukin-1 receptor antagonist (IL-1ra) in serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection technology offering high sensitivity a wide dynamic range and low sample volume needs The kit includes a 96-well plate with capture antibodies detection antibodies calibrators controls diluent read buffer and necessary reagents It shows high specificity for native human IL-1ra with no cross-reactivity and is validated for various human samples ideal for biomarker and drug discovery research
Catalog No. 50-000-81073 Supplier Meso Scale Discovery Supplier No. K156XPK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX NHP IL-1ra Assay is a multiplex immunoassay kit for measuring human interleukin-1 receptor antagonist (IL-1ra) in serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection technology offering high sensitivity a wide dynamic range and low sample volume needs The kit includes a 96-well plate with capture antibodies detection antibodies calibrators controls diluent read buffer and necessary reagents It shows high specificity for native human IL-1ra with no cross-reactivity and is validated for various human samples ideal for biomarker and drug discovery research