Learn More
Meso Scale Discovery U-PLEX NHP IL-1RA ASY384-well
SDP
Catalog No. 5000081769
Encompass
The Meso Scale Discovery U-PLEX NHP IL-1ra 384-Well Assay Kit measures human interleukin-1 receptor antagonist (IL-1ra) in serum plasma and cell culture supernatants using electrochemiluminescence (ECL) detection It offers high sensitivity broad dynamic range and precision A Meso Scale Discovery SECTOR Imager is required The kit includes a 384-well plate with IL-1ra capture antibody detection antibody buffers and calibrators Sensitivity is 0 08 pg/mL with an upper quantification limit of 2500 pg/mL
Catalog No. 50-000-81769 Supplier Meso Scale Discovery Supplier No. K256XPK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX NHP IL-1ra 384-Well Assay Kit measures human interleukin-1 receptor antagonist (IL-1ra) in serum plasma and cell culture supernatants using electrochemiluminescence (ECL) detection It offers high sensitivity broad dynamic range and precision A Meso Scale Discovery SECTOR Imager is required The kit includes a 384-well plate with IL-1ra capture antibody detection antibody buffers and calibrators Sensitivity is 0 08 pg/mL with an upper quantification limit of 2500 pg/mL