Learn More
Meso Scale Discovery U-PLEX NHP IL-1RA ASYQP
SDP
Catalog No. 5000078492
Encompass
The K156XPK-1 is a Meso Scale Discovery (MSD) electrochemiluminescence immunoassay kit designed for quantitative measurement of specific biomarkers in biological samples This multiplex assay platform utilizes MSD s proprietary MULTI-SPOT technology enabling simultaneous detection of multiple analytes from a single sample with high sensitivity and broad dynamic range The kit includes pre-coated plates detection antibodies calibrators and necessary reagents for reliable reproducible results in research applications
Catalog No. 50-000-78492 Supplier Meso Scale Discovery Supplier No. K156XPK22
The K156XPK-1 is a Meso Scale Discovery (MSD) electrochemiluminescence immunoassay kit designed for quantitative measurement of specific biomarkers in biological samples This multiplex assay platform utilizes MSD s proprietary MULTI-SPOT technology enabling simultaneous detection of multiple analytes from a single sample with high sensitivity and broad dynamic range The kit includes pre-coated plates detection antibodies calibrators and necessary reagents for reliable reproducible results in research applications