Learn More
Meso Scale Discovery U-PLEX NHP IL-2 ASY 25 PL
SDP
Catalog No. 5000080811
Encompass
The Meso Scale Discovery U-PLEX NHP IL-2 Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human non-human primate and mouse interleukin 2 (IL-2) in serum plasma and cell culture supernatants The assay uses MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results in just 3 5 hours with a lower limit of detection of 0 06 pg/mL and an upper limit of 10 000 pg/mL The kit includes a 96-well plate coated with IL-2 capture antibody detection antibody read buffer calibrator diluent wash buffer and a vial of lyophilized calibrator
Catalog No. 50-000-80811 Supplier Meso Scale Discovery Supplier No. K156TVK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX NHP IL-2 Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human non-human primate and mouse interleukin 2 (IL-2) in serum plasma and cell culture supernatants The assay uses MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results in just 3 5 hours with a lower limit of detection of 0 06 pg/mL and an upper limit of 10 000 pg/mL The kit includes a 96-well plate coated with IL-2 capture antibody detection antibody read buffer calibrator diluent wash buffer and a vial of lyophilized calibrator