Learn More
Meso Scale Discovery U-PLEX NHP IL-2 ASY
SDP
Catalog No. 5000081109
Encompass
The Meso Scale Discovery U-PLEX NHP IL-2 Assay Kit is an electrochemiluminescent immunoassay for measuring IL-2 in human non-human primate and rodent samples It uses MSD s SULFO-TAG labels and multi-array technology for sensitive quantitation with a detection range of 0 06 to 10 000 pg/mL Compatible with MSD s SECTOR Imager and Discovery Workbench software the kit includes a 96-well plate with IL-2 capture antibody detection antibody calibrator diluent wash buffer read buffer and reagents for one plate
Catalog No. 50-000-81109 Supplier Meso Scale Discovery Supplier No. K156TVK2
The Meso Scale Discovery U-PLEX NHP IL-2 Assay Kit is an electrochemiluminescent immunoassay for measuring IL-2 in human non-human primate and rodent samples It uses MSD s SULFO-TAG labels and multi-array technology for sensitive quantitation with a detection range of 0 06 to 10 000 pg/mL Compatible with MSD s SECTOR Imager and Discovery Workbench software the kit includes a 96-well plate with IL-2 capture antibody detection antibody calibrator diluent wash buffer read buffer and reagents for one plate