Learn More
Meso Scale Discovery U-PLEX NHP IL-2 ASY
SDP
Catalog No. 5000081553
Encompass
The Meso Scale Discovery U-PLEX NHP IL-2 Assay Kit is an electrochemiluminescent immunoassay for measuring human cynomolgus monkey or rhesus macaque IL-2 in serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology for sensitive detection on the MESO QuickPlex SQ 120 instrument The kit includes a plate with capture antibodies detection antibodies with ruthenium labels calibrators and necessary reagents The assay sensitivity is 0 06 pg/mL with an upper quantification limit of 1500 pg/mL for human IL-2
Catalog No. 50-000-81553 Supplier Meso Scale Discovery Supplier No. K156TVK1
The Meso Scale Discovery U-PLEX NHP IL-2 Assay Kit is an electrochemiluminescent immunoassay for measuring human cynomolgus monkey or rhesus macaque IL-2 in serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology for sensitive detection on the MESO QuickPlex SQ 120 instrument The kit includes a plate with capture antibodies detection antibodies with ruthenium labels calibrators and necessary reagents The assay sensitivity is 0 06 pg/mL with an upper quantification limit of 1500 pg/mL for human IL-2