Learn More
Meso Scale Discovery U-PLEX NHP IL-2 ASY384-well
SDP
Catalog No. 5000080006
Encompass
The Meso Scale Discovery U-PLEX NHP IL-2 384-Well Assay Kit is a multiplex immunoassay that measures natural human IL-2 in serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and rapid time to results This 384-well plate assay allows for efficient and high-throughput analysis of natural human IL-2 With a measuring range of 0 06-2500 pg/mL it is well-suited for quantifying IL-2 levels across a wide variety of research applications The kit includes all necessary reagents and plates for performing the immunoassay
Catalog No. 50-000-80006 Supplier Meso Scale Discovery Supplier No. K256TVK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX NHP IL-2 384-Well Assay Kit is a multiplex immunoassay that measures natural human IL-2 in serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and rapid time to results This 384-well plate assay allows for efficient and high-throughput analysis of natural human IL-2 With a measuring range of 0 06-2500 pg/mL it is well-suited for quantifying IL-2 levels across a wide variety of research applications The kit includes all necessary reagents and plates for performing the immunoassay