Learn More
Meso Scale Discovery U-PLEX NHP IL-22 ASY
SDP
Catalog No. 5000079778
Encompass
The Meso Scale Discovery U-PLEX NHP IL-22 Assay Kit is a multiplex immunoassay that measures natural human IL-22 in serum plasma and cell culture supernatants It uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision The assay requires 50 L or less sample volume and takes approximately 3 5 hours to complete It includes a plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent labels calibrator diluent wash buffer read buffer and a standard curve ranging from 1 22 to 10 000 pg/mL This assay allows for the accurate quantitation of natural human IL-22
Catalog No. 50-000-79778 Supplier Meso Scale Discovery Supplier No. K156WIK2
The Meso Scale Discovery U-PLEX NHP IL-22 Assay Kit is a multiplex immunoassay that measures natural human IL-22 in serum plasma and cell culture supernatants It uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision The assay requires 50 L or less sample volume and takes approximately 3 5 hours to complete It includes a plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent labels calibrator diluent wash buffer read buffer and a standard curve ranging from 1 22 to 10 000 pg/mL This assay allows for the accurate quantitation of natural human IL-22