Learn More
Meso Scale Discovery U-PLEX NHP IL-22 ASY
SDP
Catalog No. 5000079896
Encompass
The Meso Scale Discovery U-PLEX NHP IL-22 Assay Kit is an electrochemiluminescent immunoassay for measuring IL-22 in human non-human primate and rodent samples It uses MSD s SULFO-TAG labels and multi-array technology for sensitive reproducible results with low sample volumes The kit includes a 96-well plate with capture antibodies detection antibodies with SULFO-TAG calibrators and a read buffer for signal detection on MSD platforms Sensitivity is 0 3 pg/mL with an upper quantification limit of 2500 pg/mL
Catalog No. 50-000-79896 Supplier Meso Scale Discovery Supplier No. K156WIK1
The Meso Scale Discovery U-PLEX NHP IL-22 Assay Kit is an electrochemiluminescent immunoassay for measuring IL-22 in human non-human primate and rodent samples It uses MSD s SULFO-TAG labels and multi-array technology for sensitive reproducible results with low sample volumes The kit includes a 96-well plate with capture antibodies detection antibodies with SULFO-TAG calibrators and a read buffer for signal detection on MSD platforms Sensitivity is 0 3 pg/mL with an upper quantification limit of 2500 pg/mL