Learn More
Meso Scale Discovery U-PLEX NHP IL-22 ASY384-well
SDP
Supplier:  Meso Scale Discovery K256WIK2
Encompass
The Meso Scale Discovery U-PLEX NHP IL-22 384-Well Assay Kit is a 384-well multiplex assay that measures human non-human primate and rodent IL-22 in serum plasma cell culture supernatants and other biological samples It utilizes MSD s electrochemiluminescence detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements The kit includes a 384-well plate coated with capture antibodies detection antibodies and calibrators and controls needed to quantify IL-22 levels It enables efficient and accurate quantification of IL-22 biomarkers related to immune system function and inflammation
Catalog No. 50-000-79205
Add to Cart
Request a Quote